Learn More
Thermo Scientific™ Hypersil™ Silica Javelin™ Guard Columns
Product Code. 11553171 Shop All Thermo Scientific Products
Click to view available options
:
Particle Size: 5μm; 20L x 4.0mm I.D.
Product Code. 11553171
Brand: Thermo Scientific™ 30005024006
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Javelin guard columns offer convenient, easy-to-use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Silica Solid Phase
  • Excellent batch-to-batch reproducibility
  • Spherical particle with narrow particle size distribution
  • Long column lifetime
  • High performance and column efficiency
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 20 mm
Diameter (Metric) Inner 4 mm