Learn More
Thermo Scientific™ Hypersil™ Prism RP Javelin Guard Column
Product Code. 11513201 Shop All Thermo Scientific Products
Click to view available options
:
Particle Size: 5μm; 10L x 2.1mm I.D.
Product Code. 11513201
Brand: Thermo Scientific™ 32105012106
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Offered in lengths of 10 or 20 mm, and I.D. ranging from 2.1 to 4.0mm
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase RP