Learn More
Thermo Scientific™ Hypersil™ Phenyl Drop-in Guard Cartridges
Product Code. 10381105 Shop All Thermo Scientific Products
Change view
Click to view available options
Particle Size:
3 μm
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
10381105 3 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10381105 Supplier Thermo Scientific™ Supplier No. 30903014001
Estimated Shipment: 05-11-2026
Requires
Thermo Scientific Uniguard™ direct connection or stand-alone holders
Offer convenience, economy and effective protection for extending column lifetime
  • 10mm design offers maximum protection with minimal increase in retention
  • Compatible with Thermo Scientific Uniguard™ direct connection and stand-alone holders
Features of Phenyl Solid Phase
  • For the separation of certain aromatic compounds and moderately polar compounds
  • Retention characteristics similar to Hypersil MOS
  • Alternative selectivity to C8
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 3 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 4.6 mm
Stationary Phase Phenyl