Learn More
Thermo Scientific™ Hypersil GOLD™ Javelin™ Guard Columns
Product Code. 11598680 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
4 mm
Length (Metric):
10 mm
Particle Size:
3 μm
5 μm
Unit Size:
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (Metric) Particle Size Unit Size
11598680 4 mm 10 mm 5 μm Pack of 4
11593121 4 mm 10 mm 3 μm Pack of 4
11588680 2.1 mm 10 mm 5 μm Pack of 4
11583121 2.1 mm 10 mm 3 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 11598680 Lieferant Thermo Scientific™ Lieferanten-Nr. 25005014006
Big Savings for Better Separation
Use promo code 27992
Alle Werbeaktionen anzeigen
Estimated Shipment: 06-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Hypersil GOLD HPLC columns provide excellent peak shape for all analyte types
  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitvity
  • Confidence in the accuracy and quality of analytical data
Bestellinformation
Javelin guards are available in other Thermo Scientific phases. Please call your Customer Service Representative for more information.
TRUSTED_SUSTAINABILITY
Spezifikation
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 4 mm
Stationary Phase C18