Learn More
Thermo Scientific™ Hypersil GOLD™ Javelin™ Guard Columns
Product Code. 11593121 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inner:
2.1 mm
4 mm
Length (Metric):
10 mm
Particle Size:
3 μm
5 μm
Unit Size:
Pack of 4
Product Code. 11593121
Brand: Thermo Scientific™ 25003014006
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Hypersil GOLD HPLC columns provide excellent peak shape for all analyte types
  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitvity
  • Confidence in the accuracy and quality of analytical data
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your Customer Service Representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 3 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 4 mm
Stationary Phase C18