Learn More
Thermo Scientific™ Hypersil™ BDS C18 Javelin Guard Columns
Product Code. 11528710 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
4 mm
Length (Metric):
10 mm
20 mm
Particle Size:
3 μm
5 μm
Unit Size:
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (Metric) Particle Size Unit Size
11528710 2.1 mm 10 mm 5 μm Pack of 4
11553151 4 mm 10 mm 3 μm Pack of 4
11573151 3 mm 20 mm 3 μm Pack of 4
11548710 4 mm 10 mm 5 μm Pack of 4
11583151 2.1 mm 20 mm 5 μm Pack of 4
11503161 4 mm 20 mm 5 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11528710 Supplier Thermo Scientific™ Supplier No. 28105012106
 Restricted Item
Estimated Shipment: 12-11-2026
Offer convenient, easy-to-use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of the BDS C18 Solid Phase
  • Base deactivated with minimal residual silanol activity
  • Economical with exceptional reproducibility
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase C18