Learn More
Thermo Scientific™ Hypercarb™ Preparative Guard Cartridges
Product Code. 11593201 Shop All Thermo Scientific Products
Change view
Click to view available options
:
Particle Size: 5μm; 10L x 10mm I.D.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11593201 Particle Size: 5μm; 10L x 10mm I.D.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11593201 Supplier Thermo Scientific™ Supplier No. 35005019023
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use preparative guard cartridges to extend column lifetime
  • Suited to solve problem separations
  • Exceptional retention of very polar analytes
  • True orthoganality to C18
  • Separated structurally related substances
  • Excellent for high temperature applications
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 10 mm
Stationary Phase Hypercarb