Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Single-position Cell Holder
Product Code. 11881872 Shop All Thermo Scientific Products
Click to view available options
Type:
Cell Holder
Unit Size:
Each
Product Code. 11881872
Brand: Thermo Scientific™ 4010000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Single-position Cell Holder. As supplied with GENESYS 20 Spectrophotometer. Holds one 10mm square cuvette, 1/2 in. or 10mm diameter test tube.
TRUSTED_SUSTAINABILITY
Specifications
Type Cell Holder
For Use With (Equipment) Genesys™ 20 spectrophotometer
Holds One 10 mm square cuvette, 0.5 in. or 10 mm dia. test tube
Pathlength 10 mm