Learn More
Thermo Scientific™ GENESYS™ 10 UV-Vis Spectrophotometers Test Tube Holder
Shop All Thermo Scientific Products
Click to view available options
Type:
Test Tube Holder
Unit Size:
Each
Description
- Accommodates one 25 dia. × 102mmH test tube or COD reagent vial
- For GENESYS 10 spectrophotometers with six-position cell holder (holds three) or single-cell holder platform (holds one)
Specifications
Specifications
| Type | Test Tube Holder |
| For Use With (Equipment) | Genesys™ 10 UV-vis spectrophotometer |
| Holds | One 25 dia. x 102 mm H test tube or COD reagent vial |