Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Product Code. 11771151 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
4-position carousel base
GENESYS 140/150/180,BioMate160
GENESYS 30/40/50
Spectronic200,GENESYS 30/40/50
Type:
Beckman-style Cell Holder
Cylindrical Cell Holder
Disposable Microcell Holder
Hose Entry/Exit Kit
Indiv Adjustable Filter Holder
Long Path Cell Holder
Sample Holder for Film/Filter
Single Cell Sample Holder
Test Tube Cap
Test Tube and Vial Holder
Holds:
10 mm cuvette (1)
10 to 100 mm cuvette (1)
12 mm to 25 mm tubes and vials
20 to 100 mm cuvette (1)
5 - 100 mm cylindrical cells
5 to 100 mm cuvette (1)
5 to 100mm cylindrical cuvette
50 μL microcells, Uvettes
Beckman-style short microcells
Entry and exit hoses (incl)
Filter/lenses, 10 mm thick
Filter/lenses, 6 mm thick
Filters up to 10 mm thick
Single-cell cuvette, 10mm tube
Square 10 mm PL Cuvette
Tall tubes, vials (lid open)
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type Holds unitSize
11771151 GENESYS 30/40/50 Single Cell Sample Holder Single-cell cuvette, 10mm tube Each
15886085 GENESYS 140/150/180,BioMate160 Single Cell Sample Holder 10 mm cuvette (1) Each
15896085 GENESYS 140/150/180,BioMate160 Beckman-style Cell Holder Beckman-style short microcells Each
15816095 GENESYS 140/150/180,BioMate160 Long Path Cell Holder 5 to 100mm cylindrical cuvette Each
15826095 GENESYS 140/150/180,BioMate160 Sample Holder for Film/Filter Filter/lenses, 10 mm thick Each
15836095 GENESYS 140/150/180,BioMate160 Disposable Microcell Holder 50 μL microcells, Uvettes Each
15866095 GENESYS 140/150/180,BioMate160 Long Path Cell Holder 10 to 100 mm cuvette (1) Each
11881762 Spectronic200,GENESYS 30/40/50 Single Cell Sample Holder Square 10 mm PL Cuvette Each
11893744 GENESYS 30/40/50 Indiv Adjustable Filter Holder Filters up to 10 mm thick Each
15520592 GENESYS 30/40/50 Test Tube and Vial Holder 12 mm to 25 mm tubes and vials Each
15530592 GENESYS 30/40/50 Test Tube Cap Tall tubes, vials (lid open) Each
15540592 GENESYS 30/40/50 Long Path Cell Holder 20 to 100 mm cuvette (1) Each
15550592 GENESYS 30/40/50 Long Path Cell Holder 5 to 100 mm cuvette (1) Each
15570592 GENESYS 30/40/50 Sample Holder for Film/Filter Filter/lenses, 6 mm thick Each
15580592 GENESYS 30/40/50 Hose Entry/Exit Kit Entry and exit hoses (incl) Each
11751151 4-position carousel base Cylindrical Cell Holder 5 - 100 mm cylindrical cells Each
15520602 GENESYS 30/40/50 Single Cell Sample Holder 10 mm cuvette (1) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 11771151 Supplier Thermo Scientific™ Supplier No. 336028000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Simplify sampling with easy access
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
TRUSTED_SUSTAINABILITY
Specifications
Description 10 mm Square Cell Holder
Type Single Cell Sample Holder
For Use With (Equipment) GENESYS 30/40/50
Holds Single-cell cuvette, 10mm tube
Required Accessories Accessory kit, part number 840-278200, is required for us of this accessory.
Shape Square