Learn More
Thermo Scientific™ Fluophase™ RP Javelin™ Guard Columns
Product Code. 11523321 Shop All Thermo Scientific Products
Click to view available options
:
Particle Size: 5μm; 10L x 2.1mm I.D.
Product Code. 11523321
Brand: Thermo Scientific™ 82505012106
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Fluophase RP (Perfluorohexyl) Solid Phase
  • Perfluorinated phases provide unique selectivity
  • Extra retention and selectivity for halogenenated compounds
  • Excellent for taxane analysis
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase RP