Learn More
Thermo Scientific™ Buffers for MagJET™ gDNA Kit
Product Code. p-4861464 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mL
Product Type:
Concentrated Wash Buffer 1
Unit Size:
50mL
Product Code. 13515210
Brand: Thermo Scientific™ R2462
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The Thermo Scientific™ MagJET™ Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from tissue and cell cultures, bacteria and yeast, as well as from human body samples, such as buccal and urogenital swabs, urine, saliva and hair follicles. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MagJET gDNA Kit
Product Type Concentrated Wash Buffer 1
Quantity 50 mL