Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Desthiobiotin
Shop All Thermo Scientific ProductsDescription
Thermo Scientific EZ-Link Sulfo-NHS-LC-Desthiobiotin is a long-chain, amine-reactive labeling reagent whose biotin-like group is elutable from streptavidin, making it ideal for purified or cell surface protein labeling and purification.
Sulfo-NHS-LC-Desthiobiotin is a variant of biotin that is activated as a sulfo-NHS ester with a long-chain (LC) spacer arm to covalently label primary amines (-NH2) of proteins or other molecules with desthiobiotin groups. The desthiobiotin tag binds to streptavidin and other biotin-binding proteins with high specificity yet readily elutes with mild conditions (i.e., by competitive displacement with regular, free biotin). As such, this reagent is a useful alternative to Sulfo-NHS-LC-Biotin (Part No. 21335) for avidin-biotin techniques in which nondenaturing elution of the labeled proteins is desired. The negatively charged sulfo-NHS group prevents this reagent from permeating cell membranes; therefore, Sulfo-NHS-LC-Desthiobiotin can be used to confine labeling to cell surface proteins of intact cells. The No-Weigh Format (5 x 1mg resealable vials) eliminates the difficulties associated with weighing small quantities of reagent and maximizes protection of unused reagent from hydrolysis.
Highlights:
- Desthiobiotin – analog of biotin allows easy elution from streptavidin, an ideal feature for affinity purification applications
- Protein labeling – tag antibodies or proteins in purified or mixed samples to enable high recovery in pull-down assays with streptavidin beads
- Cell surface labeling – modifies only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
- Amine-reactive – reacts with primary amines (-NH2), such as the side-chain of lysines (K) or the amino-termini of polypeptides
- Soluble – charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
Desthiobiotin is a single-ring, sulfur-free analog of biotin that binds to streptavidin with nearly equal specificity but less affinity than biotin (1/Kd = 10^11 vs. 10^15M, respectively)[1-2]. Consequently, desthiobiotinylated bait proteins and their interacting partners can be eluted readily and specifically from streptavidin affinity resin using mild conditions based on competitive displacement with free biotin. For pull-down assay experiments with biological samples, this soft-release characteristic of desthiobiotin also helps to minimize co-purification of endogenous biotinylated molecules, which remain bound to streptavidin upon elution of the target protein complexes with free biotin. The modified avidin-biotin affinity system also eliminates the need to use harsh elution conditions that might disassociate complexes and/or damage the target protein or cell. Desthiobiotin-based techniques are ideal when using native or recombinant proteins that are not expressed with a fusion tag and when isolating captured proteins under native conditions, such as targeting intact cells or cell surface proteins.
Labeling with NHS Ester Reagents:
N-Hydroxysulfosuccinimide (NHS) esters of biotin are the most popular type of biotinylation reagent. NHS-activated biotins react efficiently with primary amino groups (-NH2) in alkaline buffers to form stable amide bonds. Proteins typically have several primary amines that are available as targets for labeling, including the side chain of lysine (K) residues and the N-terminus of each polypeptide. Most sulfo-NHS esters are directly water soluble but not membrane permeable. Plain NHS esters are less soluble in aqueous buffers but are membrane permeable.
Specifications
Specifications
| Product Type | EZ-Link Sulfo-NHS-LC-Desthiobiotin, No-Weigh Format |
| Quantity | 5 x 1 mg |
| Sufficient For | 5 uses to label a total of 63mg of IgG using typical conditions |
| Content And Storage | -20°C with desiccant, protect from moisture, use only fresh solutions |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.