Learn More
Thermo Scientific™ BetaSil™ C18 Javelin™ Guard Columns
Product Code. 11548720 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
Length (Metric):
10 mm
20 mm
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (Metric) unitSize
11548720 2.1 mm 10 mm Pack of 4
11553231 2.1 mm 20 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11548720 Supplier Thermo Scientific™ Supplier No. 70105012106
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Betasil C18 Solid Phase
  • Ligand on 100Å silica
  • High surface area with high bonded phase coverage
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase C18
Product Line BetaSil