Learn More
Thermo Scientific™ Aquasil™ C18 Javelin™ Guard Columns
Product Code. 11583311 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
4 mm
Length (Metric):
10 mm
20 mm
Particle Size:
3 μm
5 μm
Unit Size:
Pack of 4
Product Code. 11583311
Brand: Thermo Scientific™ 77505022106
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Offer convenient, easy-to-use protection for analytical HPLC columns
  • Direct-connection, finger-tight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Aquasil C18 Solid Phase
  • Thermo Scientific polar endcapped C18 phase
  • Alternative selectively compared to conventional C18 phases
  • Retention of polar compounds
  • Compatible with 100% aqueous mobile phases
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 20 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase C18