Learn More
Thermo Scientific™ 4-Place Swinging Bucket Rotor Adapters
For added processing versatility
67.40€ - 119.00€
Product Code | Brand | Tube Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Type | Price | Quantity & Availability | |||||
10759612
|
Thermo Scientific™
75007542 |
50 mL Tube |
73.15€
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10294434
|
Thermo Scientific™
75007545 |
7 mL Tube |
69.35€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
10698802
|
Thermo Scientific™
75007544 |
15 mL Blood Tube |
71.70€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
10076261
|
Thermo Scientific™
75007547 |
Microtube |
119.00€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
10688802
|
Thermo Scientific™
75007557 |
Falcon |
67.40€
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
10699762
|
Thermo Scientific™
75007556 |
50 mL Falcon Tube |
71.60€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
For added processing versatility
- Maximum tube size: 30 x 117mm
Order Info
Order 4-Place Swinging Bucket Rotor (75-007-591) with Round Buckets (75-007-555). Adapters are sold individually; order quantity to equip rotor. Note: 50mL conical adapter (75-007-556) requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
Specifications
Swinging bucket rotor |