Learn More
Brady™ Thermal Transfer Printable Labels, B-430
Used to identify property from components to finished products. Brady™ Thermal Transfer Printable Labels are made of clear polyester.
Brand:  Brady™ 030190
 View more versions of this product
Product Code. 11583764
  • ONLINE EXCLUSIVE PRICE
    275.00€
    231.11€ / 1 rolls
    Save 43.89€
    15% Off
Estimated Shipment: 04-12-2026
Description
  • For use with thermal transfer printers
  • Made of BradyBondz™ translucent, high adhesion polyester material
Specifications
Specifications
Autoclavable
Recommended ribbon series: R4400; R4900; R6000; R6200
None
Gloss
Polyester (B-430)
Thermal Transfer
Non-sterile
BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
-70°C
1000 Label(s) / Roll
Rectangle
76.2 x 50.8 mm (W x H)
-70°C to +100°C
0.076 mm
50.80 x 76.20 mm
3 in.
76.20 mm
50.8 mm
UL Recognized; CSA Accepted
Acrylic, Permanent
Clear
Roll
Single-side Printable
Thermal Transfer Printer Label
Asset Tracking, Barcode Labeling, Component and Equipment Identification, General and Industrial Identification, Rating Plates
1
THT-19-430-1
1000/Pk.
76.20 mm (W) x 50.80 mm (H)
-94°F to +212°F
0.003 in.
53.98 mm
2 in.
76.2 mm
2 in.
THT-19-430-1