Learn More
Brady™ Thermal Transfer Printable Labels, B-427
Product Code. 11849644
Click to view available options
Size (Metric):
12.7 x 36.5 mm (W x H)
19.05 x 23.7 mm (W x H)
19.05 x 44.45 mm (W x H)
20.3 x 36.5 mm (W x H)
20.32 x 36.5 mm (W x H)
25.4 x 25.4 mm
25.4 x 50.8 mm (W x H)
25.4 x 57.15 mm (W x H)
25.4 x 95.25 mm (W x H)
38.1 x 19.05 mm (W x H)
48.26 x 80.95 mm (W x H)
50.8 x 9.53 mm (W x H)
55.88 x 12.7 mm (W x H)
57.15 x 50.08 mm (W x H)
63.5 x 57.15 mm (W x H)
Labels per Row:
1
2
4
Quantity:
10,000/Pk.
1000/Pk.
1500/Pk.
3000/Pk.
3500/Pk.
5000/Pk.
Unit Size:
1 roll
Product Code. 11849644
Brand: Brady™ 062333
Ideal for a variety of applications. Brady™ Thermal Transfer Printable Labels are made of self-laminating vinyl
  • Available in various sizes
Specifications
Certifications/Compliance UL Recognized
Description Thermal Transfer Printable Labels
Adhesive Type Acrylic
Color White, Translucent
Finish Matte
Format Roll
Material Vinyl, Self-laminating
Print Technology Thermal Transfer
Product Type Thermal Transfer Printer Label
For Use With (Application) Laboratory Identification - Bottle, Tube and Vial
For Use With (Equipment) BBP81;i5100;i7100;PR Plus;IP Series;THT 76 mm Core Printers;i7500
Horizontal Repeat (Metric) 27.94 mm
Labels per Row 2
Min. Operating Temperature (Metric) -70°C
Model THT-116-427-5
No. per Pack 5000 Label(s) / Roll
Quantity 5000/Pk.
Shape Rectangle
Size 25.40 mm (W) x 25.40 mm (H)
Size (Metric) 25.4 x 25.4 mm
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 28.58 mm
Wire Gauge 16 AWG - 8 AWG
Dimensions (LxW) 25.40 x 25.40 mm
Width (English) 1 in.
Width (Metric) 25.4 mm
Width (English) Printable Area 25.40 mm
Width (Metric) Printable Area 25.40 mm
Height (English) 1 in.
Height (English) Printable Area 0.375 in
Height (Metric) 25.4 mm
Height (Metric) Printable Area 9.53 mm
Alternate Part Number THT-116-427-5