Learn More
Brady™ Thermal Transfer Printable Labels
Product Code. 17150501
Click to view available options
Length (Metric):
9.53 mm
12.70 mm
25.40 mm
Color:
Blue, White
Green, White
Orange, White
Pink, White
Red, White
Yellow, White
Quantity:
10,000 Labels/Roll
2500 Labels/Roll
3000 Labels/Roll
Unit Size:
Pack of 10000
Pack of 2500
Pack of 3000
Product Code. 17150501
Brand: Brady™ 306979
Only null left
Add to basket
Add to basket
Our B-494 polyester label is a reliable sample identification and traceability solution.
  • The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
  • Thanks to subsurface printing, the label's colour code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifications
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color Pink, White
Finish Gloss
Format Roll
Material Polyester
Printable Sides Front
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Cold temperature labelling, Component and equipment labelling, Laboratory labelling
For Use With (Equipment) BBP11, BBP12, PR Plus, THT 76 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -196°C
Model BPT-310-494-2.5-PK
No. per Pack 2500 Label(s) / Roll
Quantity 2500 Labels/Roll
Shape Rectangle
Size 25.40 mm (W) x 12.70 mm (H)
Temperature Range (English) -320.8°F to +158°F
Temperature Range (Metric) -196°C to +70°C
Thickness (English) 0.003 in.
Thickness (Metric) 0.0762 mm
Vertical Repeat (Metric) 15.24 mm
Dimensions (LxW) 25.40 mm (W) x 12.70 mm (H)
Length (English) 0.5 in.
Length (Metric) 12.70 mm
Width (English) 1 in.
Width (Metric) 25.40 mm
Width (English) Printable Area 25.40 mm
Height (English) 0.5 in.
Height (Metric) 12.70 mm
Alternate Part Number BPT-310-494-2.5-PK