missing translation for 'onlineSavingsMsg'
Learn More

Interscience™ Thermal Transfer Film

Product Code. 17863836
Click to view available options
Color:
Black
Unit Size:
1 roll
This item is not returnable. View return policy

Product Code. 17863836

Brand: Interscience™ 410130

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Film capable of being used with thermal transfer printing technology.

  • For use with thermal printer

Specifications

Product Type Printer Ribbon
Color Black
For Use With (Equipment) Thermal Printer
Print Technology Thermal Transfer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.