Learn More
Cole-Parmer™ Benchtop Heat Protector, Glass Ceramic
Product Code. 11817242
Click to view available options
Length (Metric):
150 mm
228 mm
300 mm
Unit Size:
Each
Product Code. 11817242
Brand: Cole-Parmer™ 00222IZ
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Keep your benchtop safe from hot labware with this Glass Ceramic Benchtop Heat Protector.
  • Plate has corner feet to prevent heat contact with benchtop.
  • Glass ceramic surface resists stains and is easy to clean.
  • Temperature range is from -392°F to +1200°F (-236°C to +649°C).
Specifications
Product Type Benchtop Heat Protector
Length (Metric) 300 mm
Width (Metric) 300 mm
Dimensions (L x W) 300 x 300 mm
Quantity 1 Ea.