Learn More
SymLine The Werner Purging Manifold
Product Code. 16257751
Change view
Click to view available options
Thread Size:
1 x GLS80 (f) to 4 x GL45 (m)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Unit Size
16257751 1 x GLS80 (f) to 4 x GL45 (m) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16257751 Supplier SymLine Supplier No. 106660
  • ONLINE EXCLUSIVE PRICE
    254.00€
    232.44€ / Pack of 1
    Save 21.56€
    8% Off
Estimated Shipment: 11-12-2026
Install up to 4 SafetyCaps on your purging bottle with The Werner - SymLine Europe's purging manifold. No dangers, no contamination - protects people, environment and HPLC system.
  • Safe and easy handling: capillaries are reliably fixed.
  • Solvent filters have enough space due to the wide GLS80 opening.
  • THE SymLine System for Solvent Safety stays permanently closed, the safety chain remains intact during the purging process.
  • PE-HD, electrostatic conductive
Specifications
For Use With (Application) HPLC / UHPLC Systems
Description Der Werner' Collector 4-ways for HPLC rinsing bottle GLS80, PE-HD electrostatic conductive, 4 x GL45 (m)
For Use With (Equipment) Purging Bottle
Type Purging Manifold
Solvent Channels 4
Material HDPE
Color Black
Thread Size 1 x GLS80 (f) to 4 x GL45 (m)