Learn More
Abeba™ The Original 7030 Shoes
Product Code. 15662488
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15662488
Brand: Abeba™ 7030/41
Animal fats, oil and fuel resistant. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather, perforated
  • Black
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB FO SRC
Color Black
Material Leather
Size 41
Sole Features Slip-resistant
Style 7030
Resistant To Animal fats, oil and fuel