Learn More
Abeba™ The Original 7021 Shoes
Product Code. 15642478
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15642478 42 1 pair
15672468 35 1 pair
15682468 36 1 pair
15692468 37 1 pair
15602478 38 1 pair
15612478 39 1 pair
15622478 40 1 pair
15632478 41 1 pair
15652478 43 1 pair
15662478 44 1 pair
15672478 45 1 pair
15682478 46 1 pair
15692478 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15642478 Supplier Abeba™ Supplier No. 7021/42
  • ONLINE EXCLUSIVE PRICE
    40.10€
    36.21€ / 1 pairs
    Save 3.89€
    9% Off
View all promotions
Estimated Shipment: 10-11-2026
Animal fats, oil and fuel resistant. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB FO SRC
  • Smooth leather white
  • Slip resistance outsole sole
  • Sanitized treated
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB FO SRC
Color White
Material Leather
Size 42
Sole Features Slip-resistant
Style 7021
Resistant To Animal fats, oil and fuel