Learn More
Thalidomide 4'-oxyacetamide-alkylC4-amine
Product Code. 18704579
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18704579 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18704579 Supplier Tocris Bioscience Supplier No. 6469/25
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
PROTACs (Proteolysis Targeting Chimeras)
Thalidomide 4'-oxyacetamide-alkylC4-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a short alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 3
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material PROTAC Toolbox
CAS 2245697-86-1
Synonym Thalidomide - linker 3
Quantity 25 mg
Molecular Formula C19H22N4O6.HCl
Formula Weight 438.86
Purity 95%