Learn More
DiscoverX TH588
Description
TH588 is a potent selective inhibitor of MTH1 (NUDT1). It can also induce DNA damage and triggers an ATM-p53-mediated death response and DNA repair in U2OS cells. Provided as 5mg of dry powder.
Specifications
Specifications
| Quantity | 5 mg |