Learn More
DiscoverX TH588
Product Code. 17312639
Encompass
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17312639 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17312639 Supplier DiscoverX Supplier No. 921325
TH588 is an antagonist for MTH1. Provided as 5mg of dry powder.
TH588 is a potent selective inhibitor of MTH1 (NUDT1). It can also induce DNA damage and triggers an ATM-p53-mediated death response and DNA repair in U2OS cells. Provided as 5mg of dry powder.
Specifications
Quantity 5 mg