missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TH 9619 disodium salt
Description
TH 9619 disodium salt is a potent and selective methylenetetrahydrofolate dehydrogenase inhibitor (MTHFD2; IC50 = 47 nM). It shows efficacity on HL-60 cell viability with EC50 of 11nM. In vitro, It shows antiproliferative efficacy in acute myeloid leukemia (AmL) cells and T-ALL Jurkat cells. In vivo, TH9619 disodium salt impairs cancer progression in a mouse disease model of AmL.
Specifications
Specifications
| Chemical Name or Material | Disodium N-[[5-[[2-(2,4-Diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)acetyl]amino]-3-fluoro-2-pyridinyl]carbonyl]-L-glutamate |
| Quantity | 50 mg |
| Target | Dehydrogenase Inhibitors |
| Molecular Formula | C17H16FN7Na2O7 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?