Learn More
Tocris Bioscience™ TH 9619 disodium salt
Description
TH 9619 disodium salt is a potent and selective methylenetetrahydrofolate dehydrogenase inhibitor (MTHFD2; IC50 = 47 nM). It shows efficacity on HL-60 cell viability with EC50 of 11nM. In vitro, It shows antiproliferative efficacy in acute myeloid leukemia (AmL) cells and T-ALL Jurkat cells. In vivo, TH9619 disodium salt impairs cancer progression in a mouse disease model of AmL.
Specifications
Specifications
| Chemical Name or Material | Disodium N-[[5-[[2-(2,4-Diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)acetyl]amino]-3-fluoro-2-pyridinyl]carbonyl]-L-glutamate |
| Quantity | 10 mg |
| Target | Dehydrogenase Inhibitors |
| Molecular Formula | C17H16FN7Na2O7 |
| Purity | 0.98 |