Learn More
Abnova™ TGFBR3 Split FISH Probe
Product Code. 16083485
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Product Code. 16083485
Brand: Abnova™ FS0051.100uL
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Specifications
Content And Storage Store at 4°C in the dark
Format Tube
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Gene transforming growth factor, beta receptor III
Gene Alias BGCAN,betaglycan
Gene Symbol TGFBR3
Gene ID (Entrez) 7049
Product Type DNA/RNA
Quantity 100 μL
Form Liquid