Learn More
Abnova™ TGFBR3 Split FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | transforming growth factor, beta receptor III |
| Gene Alias | BGCAN,betaglycan |
| Gene Symbol | TGFBR3 |
| Gene ID (Entrez) | 7049 |
| Product Type | DNA/RNA |
| Quantity | 200 μL |
| Form | Liquid |