missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems TGF-beta Premixed Magnetic Luminex™ Performance Assay

Product Code. 30001697
Change view
Click to view available options
Quantity:
2 Plex
3 Plex
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001697 2 Plex 1 set
18753517 3 Plex 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001697 Supplier R&D Systems Supplier No. FCSTM1702
  • ONLINE EXCLUSIVE PRICE
    1440.00€
    1359.75€ / 1 sets
    Save 80.25€
    5% Off
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 Plex
Species Human, Mouse, Rat, Porcine
Binding Target TGF beta 1/2/3
For Use With (Application) ELISA
Gene ID (Entrez) 7040
Synonym ARVD1, CED, DPD1, LAP, LDS5, RNHF, TGF beta1, TGFB, TGFB1, TGFB2, TGFB3, TGFbeta, TGFbeta 1, TGFbeta 2, TGFbeta 3, transforming growth factor, beta 1
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.