Learn More
Ebro Electronic™ TFX 430 Precision Thermometer
GREENER_CHOICE
Product Code. 10635603
Click to view available options
Probe Type:
Blunt
Glass Coated
Pointed
Without probe
Length (Metric):
190 mm
355 mm
Unit Size:
Each
Product Code. 10635603
Brand: Ebro Electronic™ 13405437A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Reference thermometer with exchangeble PT 100 probe
TFX 430 Model Precision Thermometer. Powered by a replaceable lithium battery, these precision thermometers are handy, watertight temperature measurement devices for a whole range of measurement and control applications in the laboratory and in industry.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012
Disposable Single-use
For Use With (Application) Applications: Laboratory, temperature monitoring, measure air, fluids, ductile substances, aggressive media and surfaces
Height (Metric) 54 mm
Includes Thermometer, sensor, calibration certificate, instruction manual
Length (Metric) 355 mm
Measurement Time 1 to 15 sec.
Model TFX 430 and TPX 130
Probe Type Blunt
Resolution 0.01°C from −100° to +199.99°C; 0.1°C for the remaining measuring range
Weight (Metric) 90 g
Product Type Precision Thermometer
Data Storage None
Max. Temperature (English) 932°F
Max. Temperature (Metric) 500°C
Min. Temperature (English) -148°F
Min. Temperature (Metric) -100°C
Temperature Range (Metric) -100°C to +500°C
Calibrated Yes
For Use With (Equipment) AG 130, AG 140, AG 150, AX 110, TPX 130, TPX 230, TPX 330
Battery Life Approx. 5 Years
Battery Type Lithium 3 V/Typ CR 2477
Accuracy ±0.05°C/±0.09°F
Display LCD
Width (Metric) 162 mm
IP Rating IP67