Learn More
Ebro Electronic™ TFX Core Thermometer Series
GREENER_CHOICE
Product Code. 10769598
Click to view available options
Probe Type:
Blunt
Fixed
Glass Coated
Pointed
Pointed, Pt 1000 sensor
Includes:
Calibration Cert., No Cable
No Cable
PTFE Cable, Calibration Cert.
Silicone Cable
Silicone Cable, Calibration Cert.
Model:
TFX 410
TFX 410-1 & TPX 200
TFX 410-1 & TPX 400-150
TFX 410-1 TPX 400
TFX 410-1 and TPX 100
TFX 410-1 and TPX 200-40
TFX 410-1 and TPX 300
TFX 410-1 and TPX 440
TFX 420 + TPX 200
TFX 420 + TPX 400
Unit Size:
Each
Product Code. 10769598
Brand: Ebro Electronic™ 13404168A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Precise and very robust thermometer with a big display
Thermometer for various applications in food, laboratory, industial use. All thermometers includes a factory calibration certificate. The thermometer measures with PT1000 temperature sensor.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485
Disposable Single-use
For Use With (Application) For laboratory, fluids, process monitoring, air, ductile substances
Height (Metric) 54 mm
Includes Silicone Cable, Calibration Cert.
Length (Metric) 355 mm
Material Stainless Steel, Plastic
Model TFX 410-1 & TPX 400-150
Probe Type Pointed
Resolution 0.1°C
Weight (Metric) 0.36 kg
Product Type Precision Core Thermometer
Data Storage None
Description With silicone cable
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (Metric) -50°C to +300°C or -50°C to +400°C
Calibrated Factory Calibrated
For Use With (Equipment) AG 130, AG 140, AG 150, AG 160, AX 100
Battery Life Approx. 5 Years
Battery Type Lithium 3 V/Typ CR 2477
Accuracy ±0.3°C
Immersion Length 120 mm
Display LCD
Width (Metric) 162 mm