Learn More
Ebro Electronic™ TFH 610 / TFH 620 Thermo-Hygrometer series
GREENER_CHOICE
Product Code. 11753509
Click to view available options
Temperature Accuracy:
±0.3°C
±0.5°C
Temperature Range (Metric):
0°C to 50°C
0°C to 60°C
Accuracy:
±2% rH
±2.5% rH
Unit Size:
Each
Product Code. 11753509
Brand: Ebro Electronic™ 13405610A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Thermo-Hygrometer for humidity and temperature measurement.
  • robust and impact resistant
  • High accuracy
  • °C/°F selectable
  • Approximately 5 years battery life time
  • Automatic deactivation
TRUSTED_SUSTAINABILITY
Specifications
Type Thermohygrometer
Humidity Range 0 to 100%
Resolution 0.1% rH
Accuracy ±2.5% rH
Temperature Range (Metric) 0°C to 50°C
Temperature Accuracy ±0.5°C
Temperature Resolution 0.1°C
Dimensions (L x W x H) 115 x 54 x 22 mm
Weight (Metric) 90 g
Includes Fixed humidy probe