Learn More
Texwipe™ Sterile Polystyrene Swabs
Product Code. 16575930 Shop All Texwipe Products
Click to view available options
Width (English) Head:
0.22 in.
Width (Metric) Head:
5.8 mm
Unit Size:
Each
Pack of 500
Product Code. 16575930
Brand: Texwipe™ STX763
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Texwipe’s Sterile Spun Polyester Swabs are constructed using 100% USP-grade polyester, securely bonded to the wooden or polystyrene handle by a water-based adhesive.
  • Environmental monitoring
  • Cleaning with compatible solutions and solvents
  • Picking up fine powders
  • Surface sampling
  • Collection and transport of bacteriological specimens
  • Specifications
    Length (English) 6.1 in.
    Product Type Swab
    Length (Metric) 155 mm
    Material Medical-grade spun polyester
    Width (English) Head 0.22 in.
    Width (Metric) Head 5.8 mm
    Handle Type Polystyrene
    Adhesive Type Water-based
    Handle Material Polystyrene
    Tip Style Large
    Tip Material Foam