Learn More
Texwipe™ Direct Thermal Cleanroom Labels
Product Code. 15591245 Shop All Texwipe Products
Change view
Click to view available options
Size (English):
1.5 x 1.5 in.
3 x 2 in.
4 x 2 in.
Unit Size:
4 rolls
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (English) Unit Size
15591245 1.5 x 1.5 in. 4 rolls
15511255 4 x 2 in. 4 rolls
15501255 3 x 2 in. 4 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15591245 Supplier Texwipe™ Supplier No. TX515D
  • ONLINE EXCLUSIVE PRICE
    694.00€
    576.02€ / 4 rolls
    Save 117.98€
    17% Off
Estimated Shipment: 15-01-2027
Designed for critical environments where direct thermal printing is used
  • Low out-gassing polypropylene made with ultra-low leachables and low particulates
  • Quick-ship pressure sensitive
  • Available with two different adhesive levels providing permanent adhesion without leaving residue
  • Tested for surface particles, cleaned and double bagged
Specifications
Adhesive Type Acrylic
Format Label
Material Polypropylene
For Use With (Application) Designed for critical environments where direct thermal printing is used
Quantity 1000/Pk.
Size (English) 1.5 x 1.5 in.
Size (Metric) 3.8 x 3.8 cm
Temperature Range (English) -20 to 140°F
Temperature Range (Metric) -29 to 60°C
Thickness (English) 3.3 mil