Specifications
| Product Type | Replacement Head |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| For Use With (Equipment) | AlphaMop |
| Color-coded | No |
| Material | Thermoplastic |