missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium tetrafluoroborate, 97%

Product Code. 11408027
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11408027

Brand: Thermo Scientific Alfa Aesar A17436.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 661-36-9 | C4H12BF4N | 160.95 g/mol

It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic reduction of 1,6-dihalohexanes by nickel(i) salen electrogenerated at glassy carbon cathodes in dimethylformamide.

Solubility
Solubility in hot water is almost transparent.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661-36-9
Molecular Formula C4H12BF4N
Molecular Weight (g/mol) 160.95
MDL Number MFCD00011745
InChI Key XWFABLFRYCHILB-UHFFFAOYSA-N
Synonym tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate
PubChem CID 12621
IUPAC Name tetramethylazanium;tetrafluoroborate
SMILES C[N+](C)(C)C.F[B-](F)(F)F

Specifications

Melting Point 417°C to 419°C
Quantity 100 g
Beilstein 3715404
Sensitivity Hygroscopic
Formula Weight 160.95
Percent Purity 97%
Chemical Name or Material Tetramethylammonium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.