Learn More
Tetrahydrozoline-d4 Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Tetrahydrozoline-d4 Hydrochloride |
| CAS | 1246814-66-3 |
| Molecular Formula | C13H12D4N2 • HCl |
| Quantity | 10 mg |
| Synonym | 4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole-d4 Hydrochloride, Murine Plus-d4, Soothe-d4, Tetryzoline-d4 hydrochloride, Tinarhinin-d4, Tizine-d4, Tyzanol-d4, Tyzanol-d4 Hydrochloride, Tyzine-d4, Tyzine-d4 Hydrochloride, Visine-d4, Visine-d4 hydrochloride, Vislin-d4, Yxin-d4 |
| InChI Formula | InChI=1S/C13H16N2.ClH/c1-2-6-11-10(4-1)5-3-7-12(11)13-14-8-9-15-13;/h1-2,4,6,12H,3,5,7-9H2,(H,14,15);1H/i8D2,9D2; |
| SMILES | [2H]C(N1)([2H])C([2H])([2H])N=C1C(CCC2)C3=C2C=CC=C3.Cl |
| IUPAC Name | 4,4,5,5-tetradeuterio-2-(1,2,3,4-tetrahydronaphthalen-1-yl)-1H-imidazole;hydrochloride |
| Molecular Weight (g/mol) | 204.313646 |
| Formula Weight | 240.1331 |