missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl isothiocyanate, 98%

Product Code. 11332327
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332327 1 g 1g
11342327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332327 Supplier Thermo Scientific Alfa Aesar Supplier No. L09878.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 36810-87-4 | C6H9NOS

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36810-87-4
Molecular Formula C6H9NOS
MDL Number MFCD00022476
Synonym 2-(Isothiocyanatomethyl)tetrahydrofuran

Specifications

Density 1.12
Boiling Point 122°C to 125°C (18 mmHg)
Refractive Index 1.535
Quantity 1 g
UN Number UN2922
Beilstein 111696
Sensitivity Moisture sensitive
Formula Weight 143.21
Percent Purity 98%
Chemical Name or Material Tetrahydrofurfuryl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.