Learn More
Tetrafluorosuccinic acid, 97%
Product Code. 11464437
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464437 1 g 1g
11474437 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11464437 Supplier Thermo Scientific Alfa Aesar Supplier No. B20462.03
Request Bulk or Custom Format
CAS: 377-38-8 | C4H2F4O4 | 190.05 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 377-38-8
Molecular Formula C4H2F4O4
Molecular Weight (g/mol) 190.05
MDL Number MFCD00042430
InChI Key YUDUFRYTKFGQCL-UHFFFAOYSA-N
PubChem CID 67833
IUPAC Name 2,2,3,3-tetrafluorobutanedioic acid
SMILES C(=O)(C(C(C(=O)O)(F)F)(F)F)O
Specifications
Melting Point ∼115°C
Density 1.4
Boiling Point 150°C (15 mmHg)
Linear Formula HO2CCF2CF2CO2H
Quantity 1 g
UN Number UN3261
Beilstein 1790197
Synonym tetrafluorosuccinic acid, perfluorosuccinic acid, butanedioic acid, tetrafluoro, succinic acid, perfluoro, succinic acid, tetrafluoro, tetrafluorobutanedioic acid, butanedioic acid, 2,2,3,3-tetrafluoro, 2,2,3,3-tetrafluorosuccinic acid, butanedioic acid,2,2,3,3-tetrafluoro, perfluorosuccinate
Formula Weight 190.05
Percent Purity 97%
Chemical Name or Material Tetrafluorosuccinic acid
Show More Show Less
compliance-icons
Product Identifier
  • Tetrafluorosuccinic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tetrafluorosuccinicacid