Learn More
Tetraethyl orthocarbonate, 97+%
Product Code. 11443428
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11443428
Brand: Thermo Scientific Alfa Aesar A17835.06
Request Bulk or Custom Format
CAS: 78-09-1 | C9H20O4 | 192.26 g/mol
Tetraethyl Orthocarbonate is used in the synthesis of chemokine receptor-5 inhibitors against HIV-1, benzobisoxazoles and in the preparation of organic semiconductors. It is also employed in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane and in the synthesis of cross linked poly(orthocarbonate)s used as organic solvent absorbent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetraethyl Orthocarbonate is used in the synthesis of chemokine receptor-5 inhibitors against HIV-1, benzobisoxazoles and in the preparation of organic semiconductors. It is also employed in the synthesis of 2,7-dimethylene-1,4,6,9-tetraoxaspiro[4,4]nonane and in the synthesis of cross linked poly(orthocarbonate)s used as organic solvent absorbent.
Solubility
Insoluble in water. Contact with water releases a flammable alcohol. Soluble in chloroform and methanol.
Notes
Moisture Sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Store under inert atmosphere and refrigerated.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78-09-1
Molecular Formula C9H20O4
Molecular Weight (g/mol) 192.26
MDL Number MFCD00009221
InChI Key CWLNAJYDRSIKJS-UHFFFAOYSA-N
Synonym tetraethyl orthocarbonate, tetraethoxymethane, triethoxymethoxy ethane, tetraethylorthocarbonate, ethane, 1,1',1,1'-methanetetrayltetrakis oxy tetrakis, orthocarbonic acid, tetraethyl ester, tetraethyi orthocarbonate, tetra ethyl orthocarbonate, 1-triethoxymethoxy ethane
PubChem CID 66213
IUPAC Name triethoxymethoxyethane
SMILES CCOC(OCC)(OCC)OCC
Specifications
Boiling Point 158°C to 161°C
Flash Point 52°C (125°F)
Quantity 5 g
UN Number UN3272
Beilstein 1747401
Formula Weight 192.26
Percent Purity ≥98%
Chemical Name or Material Tetraethyl orthocarbonate