Learn More
Tetracosanoic-d47 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Tetracosanoic-d47 Acid |
| CAS | 68060-00-4 |
| Molecular Formula | C24 D47 H O2 |
| Quantity | 0.1 g |
| Synonym | FL 88-d47, FL 88 (Fatty Acid)-d47, L 88-d47, L 88 (Fatty Acid)-d47, Lignoceric Acid-d47, Tetracosanoic-d47 acid (9 CI), Tetracosanoic-d47 acid (9 CI), Tetracosanoic-d47 acid (9 CI), Tetracosanoic-d47 acid (9 CI), Tetracosanoic-d47 acid (9 CI) |
| InChI Formula | InChI=1 S/C24H48O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26/h2-23H2,1H3,(H,25,26)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,20,20,21,21,22,22,23,23,24,24,24-heptatetracontadeuteriotetracosanoic acid |
| Molecular Weight (g/mol) | 415.93 |
| Formula Weight | 415.6604 g/mol |
| Show More |