Learn More
Tetrachloro Biphenol S, TRC
Specifications
Specifications
| Chemical Name or Material | Tetrachloro Biphenol S |
| CAS | 30609-79-1 |
| Molecular Formula | C12H6Cl4O4S |
| Quantity | 25 mg |
| Synonym | 4,4'-Sulfonylbis[2,6-dichlorophenol], NSC 2026693,3',5,5'-Tetrachloro-4,4'-dihydroxybiphenyl Sulfone, Bis(3,5-dichloro-4-hydroxyphenyl) Sulfone |
| InChI Formula | InChI=1S/C12H6Cl4O4S/c13-7-1-5(2-8(14)11(7)17)21(19,20)6-3-9(15)12(18)10(16)4-6/h1-4,17-18H |
| SMILES | Oc1c(Cl)cc(cc1Cl)S(=O)(=O)c2cc(Cl)c(O)c(Cl)c2 |
| IUPAC Name | 2,6-dichloro-4-(3,5-dichloro-4-hydroxyphenyl)sulfonylphenol |
| Molecular Weight (g/mol) | 388.05 |
| Formula Weight | 385.8741 |