missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetracaine-d6 HCl (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Tetracaine N,N-dimethyl-d6 hydrochloride |
| CAS | 80404-52-0 |
| Molecular Formula | C15 D6 H18 N2 O2 . Cl H |
| Quantity | 0.01 g |
| Synonym | Benzoic acid, 4-(butylamino)-, 2-[di(methyl-d3)amino]ethyl ester, monohydrochloride (9 CI), 2-[Di(methyl-d3)amino]ethyl 4-(butylamino)benzoate (ester) hydrochloride (1:1), Tetracaine-d6 hydrochloride, Tetracaine-D6 (N,N-dimethyl-D6) Monohydrochloride, Tetracaine-d6 HCl (N,N-dimethyl-d6) |
| InChI Formula | InChI=1 S/C15H24N2O2.ClH/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3;/h6-9,16 H,4-5,10-12H2,1-3H3;1 H/i2D3,3D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(CCOC(=O)c1ccc(NCCCC)cc1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-[bis(trideuteriomethyl)amino]ethyl 4-(butylamino)benzoate;hydrochloride |
| Molecular Weight (g/mol) | 306.8612 |
| Formula Weight | 306.1981 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction