Learn More
Tetrabutylammonium Bicarbonate, TRC
Product Code. 30423325
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423325 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30423325 Supplier Toronto Research Chemicals Supplier No. TRCT173510100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetrabutylammonium Bicarbonate
CAS 17351-62-1
Molecular Formula C16H36N • 3[CHO]
Quantity 100 mg
Synonym Tetrabutylammonium Hydrogen Carbonate, 1-Butanaminium','N,N,N-tributyl-','carbonate (1:1), Tetrabutylammonium bicarbonate
InChI Formula InChI=1S/C16H36N.CH2O3/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1(3)4/h5-16H2,1-4H3;(H2,2,3,4)/q+1;/p-1
SMILES CCCC[N+](CCCC)(CCCC)CCCC.OC([O-])=O
IUPAC Name tetrabutylammonium hydrogen carbonate
Molecular Weight (g/mol) 303.49
Formula Weight 303.27