Learn More
Tetrabromobisphenol A Diglycidyl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | Tetrabromobisphenol A Diglycidyl Ether |
| CAS | 3072-84-2 |
| Molecular Formula | C21 H20 Br4 O4 |
| Quantity | 2.5 g |
| Synonym | Tetrabromobisphenol A-Diglycidyl Ether, Propane','2,2-bis[3,5-dibromo-4-(2,3-epoxypropoxy)phenyl]- (7CI,8CI), 2,2'-[(1-Methylethylidene)bis[(2,6-dibromo-4,1-phenylene)oxymethylene]]bis[oxirane], 2,2-Bis(4-glycidyloxy-3,5-dibromophenyl)propane, 2,2',6,6'-Tetrabromobisphenol A diglycidyl ether, 2-[[2,6-Dibromo-4-[2-[3,5-dibromo-4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]methyl]oxirane, Glycidyl tetrabromodian ether, Tetrabromobisphenol A diglycidyl ether |
| InChI Formula | InChI=1S/C21H20Br4O4/c1-21(2,11-3-15(22)19(16(23)4-11)28-9-13-7-26-13)12-5-17(24)20(18(25)6-12)29-10-14-8-27-14/h3-6,13-14H,7-10H2,1-2H3 |
| SMILES | CC(C)(c1cc(Br)c(OCC2CO2)c(Br)c1)c3cc(Br)c(OCC4CO4)c(Br)c3 |
| IUPAC Name | 2-[[2,6-dibromo-4-[2-[3,5-dibromo-4-(oxiran-2-ylmethoxy)phenyl]propan-2-yl]phenoxy]methyl]oxirane |
| Molecular Weight (g/mol) | 656 |
| Formula Weight | 651.8095 |