Learn More
Testosterone Undecylenate, TRC
Specifications
Specifications
| Chemical Name or Material | Testosterone 10-Undecylenate |
| CAS | 29430-26-0 |
| Molecular Formula | C30 H46 O3 |
| Quantity | 250 mg |
| Synonym | (17β)-17-[(1-Oxo-10-undecen-1-yl)oxy]-androst-4-en-3-one, Androst-4-en-3-one','17-[(1-oxo-10-undecenyl)oxy]-','(17β)- (9CI), Testosterone','10-undecenoate (6CI,7CI,8CI), Testosterone 10-undecylenate, Testosterone undecylenate |
| InChI Formula | InChI=1S/C30H46O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h4,21,24-27H,1,5-20H2,2-3H3/t24-,25-,26-,27-,29-,30-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCCCCCCCC=C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] undec-10-enoate |
| Molecular Weight (g/mol) | 454.684 |
| Formula Weight | 454.345 |