Learn More
Testosterone Hexanoate, TRC
Product Code. 30409022
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409022 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30409022 Supplier Toronto Research Chemicals Supplier No. TRCT155125100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Testosterone caproate
CAS 10312-45-5
Molecular Formula C25 H38 O3
Quantity 100 mg
Synonym 3-Oxoandrost-4-en-17beta-yl Hexanoate, Testosterone Enantate Imp. B (EP), Testosterone Isocaproate Imp. E (EP), Androst-4-en-3-one','17-[(1-oxohexyl)oxy]-','(17β)-, Testosterone','hexanoate (6CI,7CI,8CI), (17β)-17-[(1-Oxohexyl)oxy]androst-4-en-3-one, 17β-Hydroxyandrost-4-en-3-one hexanoate, Testosterone 17-caproate, Testosterone caproate
InChI Formula InChI=1S/C25H38O3/c1-4-5-6-7-23(27)28-22-11-10-20-19-9-8-17-16-18(26)12-14-24(17,2)21(19)13-15-25(20,22)3/h16,19-22H,4-15H2,1-3H3/t19-,20-,21-,22-,24-,25-/m0/s1
SMILES CCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] hexanoate
Molecular Weight (g/mol) 386.57
Formula Weight 386.2821