missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Cypionate, TRC

Product Code. 30418550
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418550 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418550 Supplier Toronto Research Chemicals Supplier No. TRCT155100100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Testosterone Cypionate
CAS 58-20-8
Molecular Formula C27 H40 O3
Quantity 100 mg
Synonym Androst-4-en-3-one','17-(3-cyclopentyl-1-oxopropoxy)-','(17β)-, Testosterone','cyclopentanepropionate (6CI,7CI,8CI), Cyclopentanepropionic acid','ester with testosterone (8CI), 17β-Hydroxyandrost-4-en-3-one cyclopentanepropionate, 17β-Hydroxyandrost-4-en-3-one cyclopentylpropionate, DepAndro 100, DepAndro 200, Depo, Depo-testosterone, Deposteron, Depotest, Depovirin, NSC 9157, Pertestis, Testex Leo, Testodrin prolongatum, Testosterone 17β-cyclopentanepropionate, Testosterone 17β-cyclopentylpropionate, Testosterone 17β-cypionate, Testosterone cyclopentylpropionate, Testosterone cypionate, Testosterone','3-cyclopentanepropionate, Virilon
InChI Formula InChI=1S/C27H40O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h17-18,21-24H,3-16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCC5CCCC5
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 3-cyclopentylpropanoate
Molecular Weight (g/mol) 412.6
Formula Weight 412.2977
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.